Learn More
Perkin Elmer US LLC PTFE Sample Tube Insert, Pkg. 20
Catalog No. 509051884
Encompass_Preferred
Recommended for solid samples. Maximum temperature 250°C.
Catalog No. 50-905-1884 Supplier Perkin Elmer US LLC Supplier No. L4071596
Recommended for solid samples. Maximum temperature 250°C.