Learn More
Electron Microscopy Sciences PTFE Like Spray, 15oz.
SDP
Supplier:  Electron Microscopy Sciences 72619
Encompass_Preferred
A TFE release agent, dry, lubricant. No discernible transfer, no migration, no build-up. Contains: Isopropanol, Poly-TFE, 1,1-Dichlorofluoroethane and Carbon Dioxide. Contains no silicones, CFC's or Methyl Chloroform. Its working temperature is up to 140 degree F.
Catalog No. 50-949-055