Learn More
Hach Company PTFE-lined PE tubing 3/8in.
Catalog No. 5000257146 Shop All Hach Company Products
Encompass
3/8in ID PTFE-lined polyethylene intake tubing 25 ft (requires Connection Kit 2186)
Catalog No. 50-002-57146 Supplier Hach Company Supplier No. 922
3/8in ID PTFE-lined polyethylene intake tubing 25 ft (requires Connection Kit 2186)