Learn More
Swagelok PTFE LINED SS BRAIDED HOSE 1/4
Catalog No. NC1300009
Encompass
PTFE-Lined, Stainless Steel Braided Hose Assembly, 1/4 in. Stainless Steel Swagelok Tube Adapters, 58.7 in length
Catalog No. NC1300009 Supplier Swagelok Supplier No. SS4BHT60
PTFE-Lined, Stainless Steel Braided Hose Assembly, 1/4 in. Stainless Steel Swagelok Tube Adapters, 58.7 in length