Learn More
Waters Corp Waters Liner Tube, PTFE, ESI
Supplier:  Waters Corp 700000296
Encompass_Preferred
This PTFE liner tube is part of Waters' tubing and fittings, designed to ensure leak-free connections with low dead volume and minimal band spreading. These components are engineered for optimal performance when used together, emphasizing the importance of matching ferrule geometry, conical ports, and tubing diameters.
  • Ensures leak-free connections
  • Provides low dead volume
  • Facilitates minimal band spreading
  • Designed for optimal performance in integrated systems
  • Offers flexibility in system setup and maintenance through various material, length, and diameter options
  • Constructed from polytetrafluorethylene (PTFE)
Catalog No. 50-466-561