Learn More
Chemglass Life Sciences Stir Bar Kit, Magnetic, PTFE
SDP
Catalog No. 50872470
Encompass_Preferred
Stir Bar Kit, Magnetic, PTFE. Stir Bar Kit containing octagon cross-section shaped magnetic stir bars having a molded-in pivot ring manufactured with FDA-grade and USP Class VI PTFE. The PTFE coated Alnico magnet permits magnetic agitation without contamination.To Consist Of The Following: 6ea CG-2001-01, 6ea CG-2001-02, 2ea CG-2001-04, 3ea CG-2001-09, 1ea CG-2001-08, 1ea CG-2001-13, 1ea CG-2001-17
Catalog No. 50-872-470 Supplier Chemglass Life Sciences Supplier No. CG200150
Stir Bar Kit, Magnetic, PTFE. Stir Bar Kit containing octagon cross-section shaped magnetic stir bars having a molded-in pivot ring manufactured with FDA-grade and USP Class VI PTFE. The PTFE coated Alnico magnet permits magnetic agitation without contamination.To Consist Of The Following: 6ea CG-2001-01, 6ea CG-2001-02, 2ea CG-2001-04, 3ea CG-2001-09, 1ea CG-2001-08, 1ea CG-2001-13, 1ea CG-2001-17