Learn More
Electron Microscopy Sciences PTFE Magnetic Stirrer USA, 50-800 RPM
SDP
Catalog No. 50996283
Encompass_Preferred
All parts are fully encapsulated in a "PTFE" body so stirrer is totally inert to the most aggressive laboratory conditions. Features a speed display, reversible spin mode, no heat transfer to media, and stutter free motion. 24 Volt stepper motor with controlled acceleration and deceleration and stall detection. Ambient temperature range 0 – 40 degree C. PFA covered lead cable fused into body.
RPM: 50-800
Catalog No. 50-996-283 Supplier Electron Microscopy Sciences Supplier No. 6091010
All parts are fully encapsulated in a "PTFE" body so stirrer is totally inert to the most aggressive laboratory conditions. Features a speed display, reversible spin mode, no heat transfer to media, and stutter free motion. 24 Volt stepper motor with controlled acceleration and deceleration and stall detection. Ambient temperature range 0 – 40 degree C. PFA covered lead cable fused into body.
RPM: 50-800