Learn More
Sterlitech Corporation PTFE Unlaminated Membrane Filters, Hydrophilic, 0.5 Micron, 47mm, 100/Pk
SDP
Catalog No. NC1679420
Encompass_Preferred
Durable, efficient, and broadly compatible, these pure, hydrophilic PTFE membranes are ideal for use with HPLC seperations, aqueous/organic solvents, and many of your other filtration needs. Pack of 100. Pore size: 0.50 microns. Diameter: 47 mm. Advantec P/N: H050A047A.
Catalog No. NC1679420 Supplier Sterlitech Corporation Supplier No. H050A047A
Durable, efficient, and broadly compatible, these pure, hydrophilic PTFE membranes are ideal for use with HPLC seperations, aqueous/organic solvents, and many of your other filtration needs. Pack of 100. Pore size: 0.50 microns. Diameter: 47 mm. Advantec P/N: H050A047A.