Learn More
Cole-Parmer PTFE Thread tape (Plumber's), 3/4"W X 43ft L
Supplier:  Cole-Parmer 0827034
Encompass_Preferred
PTFE Plumber's Tape is used on all types of threaded fittings for passage of potable and non-potable water, most chemicals, oxygen, gas, hydraulic and high-pressure lines. Withstands harsh temperatures from -450 to 550°F (-267 to 287°C). Strong chemical resistance. Stretches 50% without breaking.
Catalog No. NC9069440