Learn More
Bellco Glass, Inc. PTFE/Red Rubber Septa,9mm
SDP
Catalog No. 501945394
Encompass_Preferred
Manufactured from red ptfe silicone. Ideal for screw thread vials. MADE IN THE USA. Autoclavable Size: 9mm Septa thickness: 0.040"
Catalog No. 50-194-5394 Supplier Bellco Glass, Inc. Supplier No. 207001400
Manufactured from red ptfe silicone. Ideal for screw thread vials. MADE IN THE USA. Autoclavable Size: 9mm Septa thickness: 0.040"