Learn More
MILLER STEPHENSON CHEMICAL COMPANY INC PTFE RELSE AGENT DRY 14OZ MIN4
SDP
Catalog No. NC9288255
Encompass_Preferred
The PTFE solids incorporated in Miller-Stephenson Dry Lubricants have a low coefficient of friction to minimize "slip stick" problems. They are extremely stable, appropriate for antistick, dry film lubrication. Their superior effectiveness provides lubrication over a range of applications for which smooth, repeated movement is essential. MS-122AD (14 oz Aerosol); Removes Organic Flux, Ionic Salts and Oil
Catalog No. NC9288255 Supplier MILLER STEPHENSON CHEMICAL COMPANY INC Supplier No. MS122AD
The PTFE solids incorporated in Miller-Stephenson Dry Lubricants have a low coefficient of friction to minimize "slip stick" problems. They are extremely stable, appropriate for antistick, dry film lubrication. Their superior effectiveness provides lubrication over a range of applications for which smooth, repeated movement is essential. MS-122AD (14 oz Aerosol); Removes Organic Flux, Ionic Salts and Oil