Learn More
Chemglass Life Sciences PTFE Sadlle O-ring, 25.4mm
SDP
Catalog No. CG196879
Encompass_Preferred
PTFE Saddle O-ring, 25.4mm. Saddle o-ring replacement for the CG1968 series of stirrer bearings. The saddle o-ring provides only glass and PTFE contact with the reaction, while allowing for the vacuum tight seal. Design of the bearing permits replacement of the saddle o-ring at a fraction of the cost of the entire bearing.
Catalog No. CG196879 Supplier Chemglass Life Sciences Supplier No. CG196879
PTFE Saddle O-ring, 25.4mm. Saddle o-ring replacement for the CG1968 series of stirrer bearings. The saddle o-ring provides only glass and PTFE contact with the reaction, while allowing for the vacuum tight seal. Design of the bearing permits replacement of the saddle o-ring at a fraction of the cost of the entire bearing.