Learn More
Sigma Aldrich Fine Chemicals Biosciences PTfE screw propeller stirr1EA
Catalog No. 502900460
Encompass
Guaranteed chemically resistant inert and leak-freeUse up to 280C (540f)Encapsulated stainless steel core for maximum rigidityPlease note The propeller and shaft are fused together.
Catalog No. 50-290-0460 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2151981EA
Guaranteed chemically resistant inert and leak-freeUse up to 280C (540f)Encapsulated stainless steel core for maximum rigidityPlease note The propeller and shaft are fused together.