Learn More
Bellco Glass, Inc. PTFE Sealing Film, 5 miL,96W
SDP
Catalog No. 501945631
Encompass_Preferred
DescriptionThis PTFE, 2mil thick adhesive free sealing film sheet with 96 well pattern (Patented) is specifically designed for robotic and high-throughput screening labs.Product DetailsSw Featured: NoGraduated: NoSilanized: NoLength Inches: 6Width Inches: 4Case Height Inches: 6Case Weight Lbs: 1Country Of Origin: USHarmonization Code: 3919.90.0000Round Bottom: NoCase Quantity: 100Material: PTFESterile: NoFormat: Sheet
Catalog No. 50-194-5631 Supplier Bellco Glass, Inc. Supplier No. 451004000
DescriptionThis PTFE, 2mil thick adhesive free sealing film sheet with 96 well pattern (Patented) is specifically designed for robotic and high-throughput screening labs.Product DetailsSw Featured: NoGraduated: NoSilanized: NoLength Inches: 6Width Inches: 4Case Height Inches: 6Case Weight Lbs: 1Country Of Origin: USHarmonization Code: 3919.90.0000Round Bottom: NoCase Quantity: 100Material: PTFESterile: NoFormat: Sheet