Learn More
Sigma Aldrich Fine Chemicals Biosciences PTfE sealing tape W 1/2 i
Catalog No. 502900432
Encompass
Eliminates the need for lubricants and vacuum greases on ground-glass joints desiccators bell jars etc. Provides a vacuum-tight seal yet will release readily even after long periods of time.
Catalog No. 50-290-0432 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z1043881PAK
Eliminates the need for lubricants and vacuum greases on ground-glass joints desiccators bell jars etc. Provides a vacuum-tight seal yet will release readily even after long periods of time.