Learn More
Ace Glass, Inc. PTFE SEALING TAPE, 1/4"
SDP
Catalog No. NC9702671
Encompass
99% pure PTFE tape used to create a leak-tight seal when connecting items such as a pressure gauge to an adapter. Wrap tape around male thread before threading into female. Thickness is .0035".
Catalog No. NC9702671 Supplier Ace Glass, Inc. Supplier No. 1412014
99% pure PTFE tape used to create a leak-tight seal when connecting items such as a pressure gauge to an adapter. Wrap tape around male thread before threading into female. Thickness is .0035".