Learn More
Bellco Glass, Inc. PTFE Sil Liner, 32mm
SDP
Catalog No. 501945756
Encompass_Preferred
Use with open top Polypropylene caps for applications requiring addition withdraw of vessel contents by syringe. Autoclavable to 140C. MADE IN THE USA Dimensions: 170x125mm
Catalog No. 50-194-5756 Supplier Bellco Glass, Inc. Supplier No. 563700038
Use with open top Polypropylene caps for applications requiring addition withdraw of vessel contents by syringe. Autoclavable to 140C. MADE IN THE USA Dimensions: 170x125mm