Learn More
Bellco Glass, Inc. PTFE Sil Liner, 45mm
SDP
Catalog No. 501945753
Encompass_Preferred
Use with open top polypropylene caps for applications requiring addition withdraw of vessel contents by syringe. Autoclavable to 140c. MADE IN THE USA Dimensions: 170x125mm
Catalog No. 50-194-5753 Supplier Bellco Glass, Inc. Supplier No. 563700030
Use with open top polypropylene caps for applications requiring addition withdraw of vessel contents by syringe. Autoclavable to 140c. MADE IN THE USA Dimensions: 170x125mm