Learn More
Chemglass Life Sciences Natural PTFE/Silicone Bonded into 9mm Blue Cap with Aperture Size 5.73
SDP
Catalog No. 501532580
Encompass_Preferred
Natural PTFE/Silicone Bonded into 9mm Blue Cap with Aperture Size 5.73. Bonded closures feature clear PTFE/Silicone liner bonded directly to the closure without the use of glue or adhesives. These bonded closures provide a cost effective, integrated, one-piece system that eliminates costly line fallout.
Catalog No. 50-153-2580 Supplier Chemglass Life Sciences Supplier No. CV3843CERT
Natural PTFE/Silicone Bonded into 9mm Blue Cap with Aperture Size 5.73. Bonded closures feature clear PTFE/Silicone liner bonded directly to the closure without the use of glue or adhesives. These bonded closures provide a cost effective, integrated, one-piece system that eliminates costly line fallout.