Learn More
Heidolph™ PTFE/Silicone Septa
SDP
Catalog No. 05403962 Shop All Heidolph North America Products
Change view
Click to view available options
Diameter (Metric):
20 mm
24.5 mm
Connection Type:
B14/23
B19/26
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Connection Type
05-403-962 20 mm B14/23
05-403-963 24.5 mm B19/26
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05-403-962 Supplier Heidolph™ Supplier No. 015880700
Add to Cart
Add to Cart
Heidolph™ PTFE/Silicone Septa for use with Carousel Plus Reaction System
  • Available in various diameters and connection types
  • For use with Carousel Plus Reaction System
Specifications
Connection Type B14/23
Material PTFE, Silicone
Type Septa
For Use With (Equipment) Carousel Plus Reaction System
Quantity 50/Pk.
Diameter (Metric) 20 mm
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov