Learn More
Bellco Glass, Inc. PTFE/Silicone Septa,9mm
SDP
Catalog No. 501945395
Encompass_Preferred
Manufactured from red ptfe silicone. Ideal for screw thread vials. MADE IN THE USA. Autoclavable Size: 9mm Septa thickness: 0.040"
Catalog No. 50-194-5395 Supplier Bellco Glass, Inc. Supplier No. 207001500
Manufactured from red ptfe silicone. Ideal for screw thread vials. MADE IN THE USA. Autoclavable Size: 9mm Septa thickness: 0.040"