Learn More
Chemglass Life Sciences Sleeve, PTFE, 24/40 Joint Size
SDP
Catalog No. CG13704
Encompass_Preferred
Sleeve, PTFE, 24/40 Joint Size. A .002in thick PTFE sleeve for standard taper joints provides a grease-free seal. Sold in packages of 3 each.
Catalog No. CG13704 Supplier Chemglass Life Sciences Supplier No. CG13704
Sleeve, PTFE, 24/40 Joint Size. A .002in thick PTFE sleeve for standard taper joints provides a grease-free seal. Sold in packages of 3 each.