Learn More
Perkin Elmer US LLC PTFE Spacers for Semi-Demountable Cells - PTFE, 0.2 mm, Qty. 3
Catalog No. 502111921
Encompass_Preferred
PerkinElmer supplies rectangular spacers made of either lead or teflon. These spacers come in a variety of thicknesseses, allowing you to customize the Universal Demountable Cell's path length to the experiment's requirements. Also available is a packet of Assorted Rectangular Spacers (L1270421) which includes two of each type of spacer. Please note: When rectangular windows and spacers are used, the Universal Demountable Cell becomes a Semi-Demountable Cell.
Catalog No. 50-211-1921 Supplier Perkin Elmer US LLC Supplier No. L1270408
PerkinElmer supplies rectangular spacers made of either lead or teflon. These spacers come in a variety of thicknesseses, allowing you to customize the Universal Demountable Cell's path length to the experiment's requirements. Also available is a packet of Assorted Rectangular Spacers (L1270421) which includes two of each type of spacer. Please note: When rectangular windows and spacers are used, the Universal Demountable Cell becomes a Semi-Demountable Cell.