Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach Company PTFE-SS Strainer 11 inch

Catalog No. 5000256998 Shop All Hach Company Products
Encompass

PTFE/stainless steel strainer 11in long 875in outside diameter

Catalog No. 50-002-56998 Supplier Hach Company Supplier No. 903
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PTFE/stainless steel strainer 11in long 875in outside diameter

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.