Learn More
Chemglass Life Sciences PTFE Stems, Buchner Funnel, 29/42 Joint Size
SDP
Catalog No. 501531106
Encompass_Preferred
PTFE tubing for 29/42 Size. For use with the 29/42 size CG-1406 series of filter funnels.
Catalog No. 50-153-1106 Supplier Chemglass Life Sciences Supplier No. CG1406E90
PTFE tubing for 29/42 Size. For use with the 29/42 size CG-1406 series of filter funnels.