Learn More
Sigma Aldrich Fine Chemicals Biosciences PTfE stirrer guides shaft1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z6493091EA
Encompass
for use with glass metal and PTfE stirrer shafts.featuresHigh level of chemical resistance Anti-whip and reduced vibration Vacuum (~5mm Hg) and pressure (3-5psi) performance No shedding Self releasing joint ring fits standard ground-glass jointMaximum recommended speeds continuous 500rpm intermittent 800rpm Unique features of the design are a permanently loaded composite PTfE/PEEK seal and a specifically fabricated glass ball-bearing for rigidity and smoothness of operation.Note PEEK has a very high level of chemical resistance with some susceptibility only to strong mineral acids.
Catalog No. 50-290-5661