Learn More
Sigma Aldrich Fine Chemicals Biosciences PTFE stirrer shaft
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z2386861EA
Encompass
Stirring shaft with steel core. Use with PTFE stirrer guides and for applications which do not require a guide. Unbreakable, scratch resistant, and usable up to 280°C.
Catalog No. NC1771752