Learn More
Sigma Aldrich Fine Chemicals Biosciences PTfE stirrer shaft shaft d1EA
Catalog No. 502900830
Encompass
Stirring shaft with steel core. Use with PTfE stirrer guides and for applications which do not require a guide. Unbreakable scratch resistant and usable up to 280C.
Catalog No. 50-290-0830 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5662921EA
Stirring shaft with steel core. Use with PTfE stirrer guides and for applications which do not require a guide. Unbreakable scratch resistant and usable up to 280C.