Learn More
Bellco Glass, Inc. PTFE Stirring Bar,41.3mm
SDP
Catalog No. 501945168
Encompass_Preferred
Molded, seamless Teflon construction ideal for procedures using all bellco magnetic stirrers. Excellent for mixing media and buffer solutions. Size: 41.3 x 9.5mm Case qty: 6
Catalog No. 50-194-5168 Supplier Bellco Glass, Inc. Supplier No. 197500158
Molded, seamless Teflon construction ideal for procedures using all bellco magnetic stirrers. Excellent for mixing media and buffer solutions. Size: 41.3 x 9.5mm Case qty: 6