Learn More
Bellco Glass, Inc. PTFE Stirring Bar,76.2mm
SDP
Catalog No. 501945170
Encompass_Preferred
Molded, seamless Teflon construction ideal for procedures using all bellco magnetic stirrers. Excellent for mixing media and buffer solutions. Size: 76.2 x 12.1mm Case qty: 3
Catalog No. 50-194-5170 Supplier Bellco Glass, Inc. Supplier No. 197500300
Molded, seamless Teflon construction ideal for procedures using all bellco magnetic stirrers. Excellent for mixing media and buffer solutions. Size: 76.2 x 12.1mm Case qty: 3