Learn More
Bellco Glass, Inc. PTFE Stirring Bar,82.6mm
SDP
Catalog No. 501945171
Encompass_Preferred
Molded, seamless Teflon construction ideal for procedures using all bellco magnetic stirrers. Excellent for mixing media and buffer solutions. Size: 82.6 x 19mm Case qty: 2
Catalog No. 50-194-5171 Supplier Bellco Glass, Inc. Supplier No. 197500314
Molded, seamless Teflon construction ideal for procedures using all bellco magnetic stirrers. Excellent for mixing media and buffer solutions. Size: 82.6 x 19mm Case qty: 2