Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Parker Super O-Lube, PTFE, Dimethyl Siloxane, Viscous Liquid, 2 oz

Catalog No. 501111745
Encompass

This Super O-Lube is a viscous, colorless, PTFE-based lubricant designed for use with any rubber polymer. It is formulated as a 100% clear dimethyl siloxane polymer, providing excellent lubrication for various sealing applications.

  • Formulated with PTFE
  • Compatible with all rubber polymers
  • Viscosity of 100,000 centistokes at 25 °C
  • Specific gravity of 0.97 at 25 °C
  • Contains no hazardous ingredients

Catalog No. 50-111-1745 Supplier MSC Supplier No. 65062986
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Super O-Lube is a viscous, colorless, PTFE-based lubricant designed for use with any rubber polymer. It is formulated as a 100% clear dimethyl siloxane polymer, providing excellent lubrication for various sealing applications.

  • Formulated with PTFE
  • Compatible with all rubber polymers
  • Viscosity of 100,000 centistokes at 25 °C
  • Specific gravity of 0.97 at 25 °C
  • Contains no hazardous ingredients

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.