Learn More
Cole-Parmer Essentials PTFE Chromatography Syringe Filter, 0.2m Pore, 13mm Dia, 100/Pk
Supplier:  Cole-Parmer 1594540
Encompass_Preferred
PTFE Syringe Filters stand up to the most aggressive solvent, acid, and base samples and are ideal for filtration of highly aggressive colloidal solutions. These filters can be used under extreme chemical or temperature conditions and are ideal for analytical and research applications where non-aqueous/organic solvent filtration is required. Maximum use temperature is 60C (140F).
Catalog No. NC1837667