Learn More
IW Tremont Co Inc LabExact Sterile PTFE Syringe Filters, 0.22um, 25mm, Hydrophobic, 100 pack
SDP
Supplier:  IW Tremont Co Inc LEIWTES10026
Encompass_Preferred
LabExact Disposable sterile PTFE hydrophobic filtration media syringe filters feature Luer-Lok and pressure fitting. Ultrasonically bonded for reliability and the filter media type is clearly inscribed on barrel. Excellent unit-to-unit and lot-to-lot reproducibility and consistency. The unique concentric funnel shaped barrel increases volume and decreases back pressure. PTFE membranes are inherently hydrophobic; they are ideal for the filtration of nonpolar liquids and gases and are very resistant to solvents as well as acids or bases and compatible with harsh chemicals. Use to filter and de-gassing chromatography solvents viscous organic-based HPLC samples. Ideal for aerosol sampling and can be used as a moisture barrier for venting applications and vacuum pump line protection. After pre-wetting w/alcohol, may be used for fine particle removal of aqueous & mild organic solutions in Ion Chromatography. Can be made more hydrophilic by flushing w/alcohol, followed by water.
Catalog No. 50-223-7730