Learn More
Cole-Parmer Cole-Parmer PTFE Tissue Grinder, Plain Plunger, 10 mL, 270 mm L
Catalog No. NC1691844 Shop All Cole-Parmer Products
Encompass_Preferred
These tissue grinders are capable of reducing cellular tissue to primary cells. Glass tubes are made of precision borosilicate glass. The pure PTFE plunger head is threaded onto a 6.5 mm stainless steel shaft. Grinding chamber clearance is 0.15 to 0.25 mm.
Catalog No. NC1691844 Supplier Cole-Parmer Supplier No. 4446803
These tissue grinders are capable of reducing cellular tissue to primary cells. Glass tubes are made of precision borosilicate glass. The pure PTFE plunger head is threaded onto a 6.5 mm stainless steel shaft. Grinding chamber clearance is 0.15 to 0.25 mm.