Learn More
Cole-Parmer Cole-Parmer PTFE Tissue Grinder, Serrated Tip Plunger, 2 mL, 230 mm L
Catalog No. NC1776568 Shop All Cole-Parmer Products
Encompass_Preferred
These tissue grinders are capable of reducing cellular tissue to primary cells. Glass tubes are made of precision borosilicate glass. The pure PTFE plunger head is threaded onto a 6.5 mm stainless steel shaft. Grinding chamber clearance is 0.15 to 0.25 mm.
Catalog No. NC1776568 Supplier Cole-Parmer Supplier No. 4446807
These tissue grinders are capable of reducing cellular tissue to primary cells. Glass tubes are made of precision borosilicate glass. The pure PTFE plunger head is threaded onto a 6.5 mm stainless steel shaft. Grinding chamber clearance is 0.15 to 0.25 mm.