Learn More
Heidolph™ PTFE Tube for Laborota 20, Hei-VAP Industrial Rotary Evaporator Glassware
SDP
Catalog No. 13900910 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Equipment):
Laborota 20, Hei-VAP Industrial Rotary Evaporator Glassware
Catalog No. For Use With (Equipment)
13-900-910 Laborota 20, Hei-VAP Industrial Rotary Evaporator Glassware
1 options
Catalog No. 13-900-910 Supplier Heidolph™ Supplier No. 036210340
PTFE Tube dispenses media manually through inlet valve into evaporating flask.
Included with Laborota 20, Hei-VAP Industrial Rotary Evaporator
Specifications
Material PTFE
Type Tubing
For Use With (Equipment) Laborota 20, Hei-VAP Industrial Rotary Evaporator Glassware
Dimensions (L x W) 6.5 x 0.5 in.
Diameter (English) 0.3 in.
Diameter (Metric) 7.5 mm
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov