Learn More
IDEX Health & Science Upchurch PTFE TUBING .010X.062X50'
Catalog No. NC1919808
Encompass_Preferred
Idex FEP Tubing, Diameter Inner: 0.25 mm, 0.01 in., Overall Length: 152.4 m, 50 ft., Natural, Diameter Outer: 0.0625 in., 0.158 cm, DuPont FEP, Certifications/Compliance: ROHS, For Use With: For General, Low Pressure AppliCatIons, Maximum Pressure: 1000 to 3000 psi
Catalog No. NC1919808 Supplier IDEX Health & Science Upchurch Supplier No. 1527L
Idex FEP Tubing, Diameter Inner: 0.25 mm, 0.01 in., Overall Length: 152.4 m, 50 ft., Natural, Diameter Outer: 0.0625 in., 0.158 cm, DuPont FEP, Certifications/Compliance: ROHS, For Use With: For General, Low Pressure AppliCatIons, Maximum Pressure: 1000 to 3000 psi