Learn More
IDEX Health & Science Upchurch PTFE TUBING .030X.062X100'
Catalog No. NC1919806
Encompass_Preferred
Idex FEP Chromatography Tubing: 100 ft., Natural, Diameter Inner: 0.03 in., Diameter Outer: 0.06 in., 0.15 cm, FEP/PTFE, Autoclavable: Autoclavable, Overall Length: 100 ft., 30.4 m, FEP Tubing, Certifications/Compliance: ROHS
Catalog No. NC1919806 Supplier IDEX Health & Science Upchurch Supplier No. 1520XL
Idex FEP Chromatography Tubing: 100 ft., Natural, Diameter Inner: 0.03 in., Diameter Outer: 0.06 in., 0.15 cm, FEP/PTFE, Autoclavable: Autoclavable, Overall Length: 100 ft., 30.4 m, FEP Tubing, Certifications/Compliance: ROHS