Learn More
IDEX Health & Science Upchurch PTFE TUBING .062X.125X50'
Catalog No. NC1919802
Encompass_Preferred
Idex FEP Chromatography Tubing: 50 ft., Natural, Diameter Inner: 0.062 in., Diameter Outer: 0.12 in., 0.3 cm, Autoclavable: Autoclavable, Overall Length: 50 ft., 15.2 m, FEP Tubing, Certifications/Compliance: ROHS
Catalog No. NC1919802 Supplier IDEX Health & Science Upchurch Supplier No. 1521L
Idex FEP Chromatography Tubing: 50 ft., Natural, Diameter Inner: 0.062 in., Diameter Outer: 0.12 in., 0.3 cm, Autoclavable: Autoclavable, Overall Length: 50 ft., 15.2 m, FEP Tubing, Certifications/Compliance: ROHS