Learn More
Chemglass Life Sciences Tubing, PTFE (FEP), 0.063in ID x 0.125in OD, 10' Lgth
SDP
Catalog No. CG116402
Encompass_Preferred
Tubing, PTFE (FEP), 0.063in ID x 0.125in OD, 10' Lgth. Chemically inert and suitable for low pressure applications, FEP tubing is recommended for all connections in the high performance, low pressure liquid chromatography system. The tubing is extruded with an outer diameter tolerance of +/- .001in. The 1/8in tubing can withstand pressure up to 500 psi. SUPPLIED IN LENGTHS OF 10 FEET.
Catalog No. CG116402 Supplier Chemglass Life Sciences Supplier No. CG116402
Tubing, PTFE (FEP), 0.063in ID x 0.125in OD, 10' Lgth. Chemically inert and suitable for low pressure applications, FEP tubing is recommended for all connections in the high performance, low pressure liquid chromatography system. The tubing is extruded with an outer diameter tolerance of +/- .001in. The 1/8in tubing can withstand pressure up to 500 psi. SUPPLIED IN LENGTHS OF 10 FEET.