Learn More
Chemglass Life Sciences FEP Tubing; 1/2" OD x 3/8" ID x 10ft Long
SDP
Catalog No. CG197503
Encompass_Preferred
FEP Tubing; 1/2" OD x 3/8" ID x 10ft Long. EP heavy wall tubing for use with Chemglass process reactors. Used to connect components having 1/4", 1/2", or 3/4"OD glass connections.
Catalog No. CG197503 Supplier Chemglass Life Sciences Supplier No. CG197503
FEP Tubing; 1/2" OD x 3/8" ID x 10ft Long. EP heavy wall tubing for use with Chemglass process reactors. Used to connect components having 1/4", 1/2", or 3/4"OD glass connections.