Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUCHI PTFE Tubing
SDP

Catalog No. 05405019 Shop All BUCHI Products
Change view
Click to view available options
Type:
Feeding Hose
Vacuum Hose
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
05-405-019 Vacuum Hose
05-405-020 Feeding Hose
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05-405-019 Supplier BUCHI Supplier No. 027277
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with industrial Rotavapor Rotary Evaporators

  • PTFE Tubing
  • For use with Rotavapor Rotary Evaporators

Specifications

Color White
Material PTFE
Type Vacuum Hose
For Use With (Application) Introducing Solvent, Back Feeding
For Use With (Equipment) Vacuum Pump
Diameter (Metric) 8/10 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.