Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Tubing, PVC, 19 mm x 12 mm, 1 m

Supplier:  Waters Corp 6435001S1

Encompass_Preferred

Tubing, PVC, 19 mm x 12 mm, 1 m

Catalog No. 50-792-289


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.