Learn More
Chemglass Life Sciences FEP Tubing; 3/4" OD x 5/8" ID x 20ft Long
SDP
Catalog No. CG197506
Encompass_Preferred
FEP Tubing; 3/4" OD x 5/8" ID x 20ft Long. EP heavy wall tubing for use with Chemglass process reactors. Used to connect components having 1/4", 1/2", or 3/4"OD glass connections.
Catalog No. CG197506 Supplier Chemglass Life Sciences Supplier No. CG197506
FEP Tubing; 3/4" OD x 5/8" ID x 20ft Long. EP heavy wall tubing for use with Chemglass process reactors. Used to connect components having 1/4", 1/2", or 3/4"OD glass connections.