Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUCHI PTFE Tubing, 460 mm
SDP

Catalog No. 05405012 Shop All BUCHI Products
Change view
Click to view available options
For Use With (Equipment):
Rotavapor
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
05-405-012 Rotavapor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-405-012 Supplier BUCHI Supplier No. 000643
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

To introduce solvent into the evaporating flask during distillation.

  • Transparent PTFE Hose
  • 4.7 x 3.9 mm

Specifications

Material PTFE
Type Hose
For Use With (Application) Solvent Introduction, Back Feeding
For Use With (Equipment) Rotavapor
Length (Metric) 460 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.