Learn More
Perkin Elmer US LLC 1.0 mm I.D. PTFE Liquid Rinse Feed Tube, 700 mm
Catalog No. 509042314
Encompass_Preferred
The tubing assembly is compatible with the AS9x, and S10 autosamplers as well as the FIAS and FIMS instruments. Other flow injection PTFE tubing assemblies are also available B0191058 (110 mm length) and B0198097 (300 mm length) and come with blue screw fittings at both ends.
Catalog No. 50-904-2314 Supplier Perkin Elmer US LLC Supplier No. B0191059
The tubing assembly is compatible with the AS9x, and S10 autosamplers as well as the FIAS and FIMS instruments. Other flow injection PTFE tubing assemblies are also available B0191058 (110 mm length) and B0198097 (300 mm length) and come with blue screw fittings at both ends.