Learn More
Perkin Elmer US LLC PTFE Tubing for GemTip/GemCone Nebulizers, 1 ft.
Catalog No. NC9500585
Encompass_Preferred
This tubing, with 0.81 mm I.D., is used in the GemClean Cross-Flow II Nebulizer (N8120516), Universal GemTip Plastic Body Nebulizer Complete Assembly for AAnalyst (N0370394), High-Sensitivity GemTip Plastic Body Nebulizer Complete Assembly for AAnalyst (N0370393), GemCone Low-Flow Nebulizer (N0770358), GemCone High Dissolved Solids Nebulizer (N0690670), GemTip Cross-Flow II Nebulizer for Optima 8x00 and Avio 200/500 (N0780546), and the GemTip Cross-Flow II Nebulizer for Optima 2x00/4x00/5x00/7x00 (N0770546).
Catalog No. NC9500585 Supplier Perkin Elmer US LLC Supplier No. 09985708
This tubing, with 0.81 mm I.D., is used in the GemClean Cross-Flow II Nebulizer (N8120516), Universal GemTip Plastic Body Nebulizer Complete Assembly for AAnalyst (N0370394), High-Sensitivity GemTip Plastic Body Nebulizer Complete Assembly for AAnalyst (N0370393), GemCone Low-Flow Nebulizer (N0770358), GemCone High Dissolved Solids Nebulizer (N0690670), GemTip Cross-Flow II Nebulizer for Optima 8x00 and Avio 200/500 (N0780546), and the GemTip Cross-Flow II Nebulizer for Optima 2x00/4x00/5x00/7x00 (N0770546).