Learn More
BURKLE INC PTFE-valve, two-way, Ø 0.27 in. (6,8 mm), NW 0.12 in. (3 mm), autoklav.
Catalog No. 5000026000
Encompass_Preferred
Two-way valves made of solid PTFE with hose nozzles. Resistant to chemicals. Very easy connection thanks to its excellent antifriction properties. Blue handles made of PP.
Catalog No. 50-000-26000 Supplier BURKLE INC Supplier No. 86100012
Two-way valves made of solid PTFE with hose nozzles. Resistant to chemicals. Very easy connection thanks to its excellent antifriction properties. Blue handles made of PP.