Learn More
BURKLE INC PTFE-valve, two-way, Ø 0.39 in. (10 mm), NW 0.16 in. (4 mm), autoklav.
Catalog No. 5000026014
Encompass_Preferred
Two-way valves made of solid PTFE with hose nozzles. Resistant to chemicals. Very easy connection thanks to its excellent antifriction properties. Blue handles made of PP.
Catalog No. 50-000-26014 Supplier BURKLE INC Supplier No. 86100015
Two-way valves made of solid PTFE with hose nozzles. Resistant to chemicals. Very easy connection thanks to its excellent antifriction properties. Blue handles made of PP.